Boc-β-chloro-Ala-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Boc-β-chloro-Ala-OH
Formula:C₈H₁₄ClNO₄
MW:223.653 g/mol
MDL Number:MFCD00672318
CAS Number:71404-98-3
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 223.653 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought