Boc-D-Arg(Pbf)-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Nα-Boc-Nomega-(2,2,4,6,7-pentamethyl-2
Formula:C₂₄H₃₈N₄O₇S
MDL Number:MFCD00236822
CAS Number:186698-61-3
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 526,649 g/mol
:Boc-D-Arg(Pbf)-OH
Storage Temperature:Freezer
Customers Who Bought This Also Bought