Boc-D-asp-Otbu
Supplier: Bachem Americas
Certificates
About this item
Synonyms: (R)-4-(tert-Butoxy)-3-((tert-butoxycarbonyl)amino)-4-oxobutanoic acid
Formula:C₁₃H₂₃NO₆
MW:289.328 g/mol
MDL Number:MFCD00236827
CAS Number:77004-75-2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 289.328 g/mol
:Boc-D-asp-Otbu
Storage Temperature:Refrigerator
Customers Who Bought This Also Bought