BG-β-Fme
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Boc-Glu-OFm
CAS Number:133906-29-3
:MFCD00151872
Formula:C₂₄H₂₇NO₆
MW:425.481 g/mol
MDL Number:MFCD00076934
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 425.481 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought