H-Lys(Boc)-NH₂·HCl
Supplier: Bachem Americas
Certificates
About this item
Synonyms: tert-Butyl (S)-(5
Formula:C₁₁H₂₄ClN₃O₃
MW:281.781 g/mol
MDL Number:MFCD00153446
CAS Number:112803-72-2
:H-Lys(Boc)-NH₂·HCl
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 281.781 g/mol
Storage Temperature:Freezer
Specification Test Results
- H-Lys(Boc)-NH₂·HCl:
Specification Test Results
Customers Who Bought This Also Bought