Boc-11-Aun-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: 11-[(tert-Butoxycarbonyl)amino]undecanoic acid,11-(Boc-amino)-undecanoic acid,11-(Boc-amino)undecanoic acid,Boc-11-Aun-OH
Formula:C₁₆H₃₁NO₄
MW:301.427 g/mol
MDL Number:MFCD00076902
CAS Number:10436-25-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 301.427 g/mol
:Boc-11-Aun-OH
Storage Temperature:Refrigerator
Customers Who Bought This Also Bought