Boc-ARG(Boc)2-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Boc-Arg(Boc)₂-OH
CAS Number:97745-69-2
:MFCD00270489
Formula:C₂₁H₃₈N₄O₈
MW:474.555 g/mol
MDL Number:MFCD00236819
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 474.555 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought