Boc-Asp(Ofm)-OH
Catalog #: A-2930.0005BA
Supplier: Bachem Americas
CAS Number: 117014-32-1
Specifications
- Size:5 g
- Cat. no.:A-2930.0005BA
- Supplier No.:A-2930.0005
Specifications
About this item
:Boc-Asp(Ofm)-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 411,454 g/mol
Formula:C₂₃H₂₅NO₆
Storage Temperature:Freezer
MDL Number:MFCD00076916
CAS Number:117014-32-1
Customers Who Bought This Also Bought