Nα-Boc-Nε-trifluoroacetyl-L-lysine
Supplier: Bachem Americas
Certificates
About this item
Synonyms: N2-Boc-N6-(2,2
Formula:C₁₃H₂₁F₃N₂O₅
MW:342.315 g/mol
MDL Number:MFCD00037104
CAS Number:16965-06-3
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 342.315 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought