Boc-L-Arg(Z)2-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Boc-Arg(Z)2-OH
Formula:C₂₇H₃₄N₄O₈
MW:542.6 g/mol
MDL Number:MFCD00236824
CAS Number:51219-19-3
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 542.6 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought