Boc-β-cyano-D-Ala-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Boc-β-cyano-D-Ala-OH
Formula:C₉H₁₄N₂O₄
MW:214.221 g/mol
MDL Number:MFCD00236830
CAS Number:184685-17-4
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 214.221 g/mol
:Boc-β-cyano-D-Ala-OH
Storage Temperature:Freezer
Customers Who Bought This Also Bought