Boc-Asp(OBzl)-OSu
Catalog #: A-1255.0025BA
Supplier: Bachem Americas
CAS Number: 13798-75-9
Specifications
- Size:25 g
- Cat. no.:A-1255.0025BA
- Supplier No.:A-1255.0025
Specifications
About this item
:Boc-Asp(OBzl)-OSu
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 420,418 g/mol
Formula:C₂₀H₂₄N₂O₈
MW:420.418 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00037912
CAS Number:13798-75-9
Customers Who Bought This Also Bought