Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride
Formula:C₁₁H₂₂N₄O₄·HCl
MW:310.78 g/mol
MDL Number:MFCD00063594
CAS Number:35897-34-8
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 310.78 g/mol
:Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride
Storage Temperature:Freezer
Customers Who Bought This Also Bought