Pentagastrin
Supplier: Bachem Americas
Certificates
About this item
Formula:C₃₇H₄₉N₇O₉S
MDL Number:MFCD00076515
CAS Number:5534-95-2
:Pentagastrin
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 767.899 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought