Succinimido (S)-2-[(tert-butoxycarbonyl)amino]propionate
Supplier: Bachem Americas
Certificates
About this item
Synonyms: N-Boc-L-alanine N-succinimidyl ester
Formula:C₁₂H₁₈N₂O₆
MW:286.3 g/mol
MDL Number:MFCD00065974
CAS Number:3392-05-0
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 286.3 g/mol
:Succinimido (S)-2-[(tert-butoxycarbonyl)amino]propionate
Storage Temperature:Freezer
EINECS:222-229-7
Customers Who Bought This Also Bought