Boc-Lys(boc)-OSu
Supplier: Bachem Americas
Certificates
About this item
Synonyms: (S)-2,5-Dioxopyrrolidin-1-yl 2,6-bis((tert-butoxycarbonyl)amino)hexanoate,Nα
Formula:C₂₀H₃₃N₃O₈
MW:443.497 g/mol
MDL Number:MFCD00057898
CAS Number:30189-36-7
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 443.497 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought