Sodium Standard Solutions, (NIST), Hach
Supplier: Hach
Certificates
About this item
For accuracy checks and instrument calibrations in sodium determinations.
Caution: 100 mg/L = 100 µg/mL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.