Order Entry
ContactUsLinkComponent
Free Chlorine Test Kit, Model CN-70F, Hach
Free Chlorine Test Kit, Model CN-70F, Hach
Catalog #: CA11014-568
Supplier:  Hach
CAS Number:  
FREE CHLORINE TEST KIT-CN-70F
Free Chlorine Test Kit, Model CN-70F, Hach
Catalog #: CA11014-568
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    DPD (N,N-diethyl-p-phenylenediamine)
  • Description:
    Free Chlorine Test Kit, Model CN-70F
  • Method:
    Color disc / DPD
  • Case style:
    D
  • Increments:
    0.1
  • Cat. no.:
    CA11014-568
  • Range:
    0.02 - 0.7 mg/l, 0 - 3.4 mg/l Cl₂
  • Supplier No.:
    1454201
  • No. of Tests:
    200

Specifications

About this item

Simple, low-cost color disc method is the most accurate visual test kit method.

Quickly and accurately measure free chlorine levels in the field or in the lab

Quickly and accurately measure free chlorine levels in the field or in the lab

*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.

Customers Who Bought This Also Bought