ContactUsLinkComponent
Laboratory CO₂ Incubators, Air Jacketed/IR Sensor, Shel Lab, Sheldon
Laboratory CO₂ Incubators, Air Jacketed/IR Sensor, Shel Lab, Sheldon
Certificates
Laboratory CO₂ Incubators, Air Jacketed/IR Sensor, Shel Lab, Sheldon
Laboratory CO₂ Incubators, Air Jacketed/IR Sensor, Shel Lab, Sheldon
Certificates
Specifications
  • Catalog No:
  • 89409-308
  • 89409-306
  • 89409-304
  • CO₂ range:
  • 0 - 20%
  • 0 - 20%
  • 0 - 20%
  • Temperature uniformity:
  • +/-0.5°C at 37°C
  • +/-0.5°C at 37°C
  • +/-0.5°C at 37°C
  • Temperature range:
  • Ambient +8…60 °C
  • Ambient +8…60 °C
  • Ambient +8…60 °C
  • Recovery Time:
  • < 5 minutes (CO2 recovery rate)
  • < 5 minutes (CO2 recovery rate)
  • < 5 minutes (CO2 recovery rate)
  • No. of outlets:
  • Single One Amp
  • Single One Amp
  • Single One Amp