Specifications
- Chemical method:Cold periodate
- Description:Manganese Test Kit, Model MN-5
- Model:MN-5
- Method:Color disc/Cold periodate
- Case style:D
- Cat. no.:CA11014-452
- Range:0.1 - 3.0 mg/l Mn
- Supplier No.:146700
- No. of Tests:100
Specifications
About this item
Manganese causes a bitter taste in water, and at concentrations above 0.1 mg/L, it causes objectionable stains on laundry and plumbing fixtures.
-Simple, low-cost color disc method is the most accurate visual test kit method
-Achieve fast results with an easy-to-carry test kit
- Simple, lowcost color disc method is the most accurate visual test kit method
- Achieve fast results with an easytocarry test kit
Manganese control also is important in the beverage, paper, and textile industries, and in dye production and food processing. An economical solution, this kit uses the simple, accurate color disc method.