Adapter, Rotary Evaporator to Freeze Drying Vessel, Ace Glass
Catalog #: 89055-794
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Adapter, Rotary Evaporator to Freeze Drying Vessel
- Cat. no.:89055-794
- ST Joint Size:24/40
- Thread Size:#25 Ace-Thread
- Supplier No.:6696-70
Specifications
About this item
By using this adapter, preliminary evaporation may be done on a rotary evaporator, then vessels can be transferred directly to freeze drying manifold
No need to transfer contents of flask. Joint is [ST] 24/40 with a 2 mm hole mid length for relieving the vacuum gently. The Ace-Thred connection is #25. For nylon bushing (with FETFE O-ring), see 89059-226. Fabricated from borosilicate glass.
Customers Who Bought This Also Bought