Specifications
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Description:Total Chlorine Color Disc Test Kit, Model CN-66T
- Model:CN-66T
- Method:Color disc/DPD
- Case style:B
- Cat. no.:CA97026-860
- Range:0 - 3.4 mg/l Cl₂
- Supplier No.:223103
- No. of Tests:100
Specifications
About this item
One of Hach's most popular test kits, it uses powder DPD reagent that reacts with chlorine more quickly than tablet-form DPD, giving more accurate results.
-One of Hach's most popular test kits
-Easy to use color disc method
-Quickly and accurately measure total chlorine lev+G74els in the field or in the lab
- One of Hach's most popular test kits
- Easy to use color disc method
- Quickly and accurately measure total chlorine lev+G74els in the field or in the lab
Powder DPD also has a considerable advantage over orthotolidine, a hazardous substance sometimes used as a chlorine test reagent
*mg/L unless otherwise noted, ppb = μg/L, ppm = mg/L.; gpg = grains per gallon; 1 gpg = 17.1 mg/L or 17.1 ppm.
The simple, low-cost color disc method is the most accurate visual test kit method. *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.