ContactUsLinkComponent
Free Chlorine Test Strips, 0-600 mg/L, Hach
Free Chlorine Test Strips, 0-600 mg/L, Hach
Catalog #: CA2890200
Supplier:  Hach
CAS Number:  
Free Chlorine Test Strips
Free Chlorine Test Strips, 0-600 mg/L, Hach
Catalog #: CA2890200
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Free Chlorine Test Strips, 0-600 mg/L
  • Increments:
    Steps: 0, 25, 50, 100, 200, 400, 600
  • Cat. no.:
    CA2890200
  • Range:
    0 - 600 mg/l Cl₂
  • Supplier No.:
    2890200
  • No. of Tests:
    100
Specifications
About this item
These test strips are manufactured with a unique reagent pad at the tip, using proven chemistries based on standard reference methods.
-Ideal for anyone performing semi-quantitative pre-screenings, spot-checks, or field tests
-Simplify testing and get results in less than 1 minute, in the field or lab
-Eliminate chemical handling and clean-up
-Reduce guesswork by screening samples
  • Ideal for anyone performing semiquantitative prescreenings, spotchecks, or field tests
  • Simplify testing and get results in less than 1 minute, in the field or lab
  • Eliminate chemical handling and cleanup
  • Reduce guesswork by screening samples
Just dip the strip, wait until color development occurs, and compare the color of the reacted pad to the chart printed on the bottle
*mg/L unless otherwise noted; ppb = μg/L; ppm = mg/L; gpg = grains per gallon; 1 gpg = 17.1 mg/L or 17.1 ppm.
*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Customers Who Bought This Also Bought