About this item
Type 2 sound meter with one-button operation.
The Sper Scientific Sound Level Pen features a type 2 internal calibrator, one-button operation, low cost, auto-ranging and max hold in a mini size.
The Sper Scientific Sound Level Pen is the smallest, lightest, easiest to carry sound meter in the world. Despite its small size, the Sound Level Pen is a full function type 2 (Meets ANSI S1.4 Type 2 and IEC61672-1 Class 2) sound meter perfect for testing OSHA requirements and most other sound meter applications. Auto-ranging with simple one-button operation. Press once to begin reading sound levels. Press the button a second time and the display holds the maximum reading, updating as new maximums are reached. Hold the button for 2 seconds to turn the unit off. Measures the 32–130 dB range in the "A" scale, with an accuracy of ±1.5 dB. The display provides 0.1 dB resolution, under range, over range and low battery indicators. The Sound Level Pen calibrates electronically to its own internal oscillator (94dB at 1kHz) or external 2 Pt. Acoustical Calibrator 850016.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 97011-816
- 97011-814
- Dimensions:
- 206 x 42 x 25 mm (8 x 1.5 x 1")
- 206 x 42 x 25 mm (8 x 1.5 x 1")
- Weight:
- 100 g (3.5 oz.), with batteries
- 100 g (3.5 oz.), with batteries