Specifications
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Description:DPD Free Chlorine Reagent Powder Pillows
- Size:5 mL
- Method:DPD
- Cat. no.:CA21978-46
- Range:up to 3.5 mg/L Cl₂
- Supplier No.:21978-46
- No. of Tests:50
Specifications
About this item
For determination of free chlorine with Hach's Chlorine Visual Test Kits
Sample Size: 5 mL. Range: up to 3.5 mg/L Cl2. *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.