To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Chemical method:Thiosulfate
- Description:Chlorine (Total) Test Kit, Model CN-65
- Model:CN-65
- Method:Drop count titration/ Thiosulfate
- Case style:D
- Increments:smallest steps - 1
- Cat. no.:CA11014-670
- Range:1 - 20 mg/l, 0.2 - 4 mg/l
- Supplier No.:225401
- No. of Tests:100
Specifications
About this item
The most widely used disinfectant for drinking water, chlorine is also important for sanitizing swimming pools, cooling towers, other industrial equipment, and in the treatment of municipal wastewater.
-Utilizes drop count titration - a simple, economical titration method
-Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
- Utilizes drop count titration a simple, economical titration method
- Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Its measurement and control are vital for both safety and economic reasons. *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.