Water Resistant Stopwatch, Sper Scientific
Supplier: Sper Scientific
Certificates
About this item
Water resistant digital stopwatch.
- Split interval function
- LCD display
Counts up to 24 hours in ¹/₁₀₀th second resolution for the first 30 minutes with a 1 second resolution thereafter. Also has a 12/24 hour alarm clock, calendar and an hourly chime option.
Certifications: N.I.S.T. traceable certificate of compliance for 97035-842.
Ordering information: Five year warranty.
Customers Who Bought This Also Bought