Specifications
- Chemical method:Thiosulfate
- Description:Chlorine (Total) Test Kit, Model CN-21P
- Model:CN-21P
- Method:Drop count titration/ Thiosulfate
- Case style:B
- Cat. no.:CA24444-00
- Range:10 - 200 mg/l
- Supplier No.:24444-00
- No. of Tests:100
Specifications
About this item
Chlorine Test Kits are manufactured to ensure compliance with stringent quality control standards.
This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
- Simple, economical titration method
- Contains all required reagents, solutions, and glassware
Using the Drop Count Titration method, the CN-21P Chlorine Test Kit is used to determine high concentrations of chlorine in sanitizing applications
This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
10-200 mg/L.
**mg/L unless otherwise noted, ppb = μg/L, ppm = mg/L; gpg = grains per gallon; 1 gpg = 17.1 mg/L or 17.1 ppm. 1EPA-accepted for wastewater. Check with your regulatory agency.