About this item
Monitor visible light levels anywhere with a fast, stable, and accurate response
The Certified Light Meter Lux/FC (97035-930) includes a N.I.S.T. traceable certificate of calibration. Each unit is also supplied with a 5-year warranty.
Results are easily read on the extra-large 38.1mm (1½") LCD with bar graph, low battery, and over range indications. The color and cosine corrected sensor meets C.I.E. photopic spectrum. The unit also features an RS232 computer output, min./max., auto power off, external zero adjustment, hold function, and a tripod back. The Lux/Foot-Candle light meter comes ready to use with a 9V battery, instructions, soft padded case with shoulder strap, and a detachable sensor (44" cord included) with cover for simple replacement or repair.
Specifications
- Catalog No:
- 97035-928
- 97035-930
- Weight:
- 280 g (10 oz.)
- 280 g (10 oz.)
- Dimensions:
- 200 x 68 x 30 mm (77/8 x 25/8 x 11/8")
- 200 x 68 x 30 mm (77/8 x 25/8 x 11/8")