Order Entry
ContactUsLinkComponent
Vibration Isolation Table, Labconco®
Vibration Isolation Table, Labconco®
Catalog #: 82000-984
Supplier:  Labconco
Product available on GSA Advantage®
Vibration Isolation Table
Vibration Isolation Table, Labconco®
Catalog #: 82000-984
Supplier:  Labconco
Product available on GSA Advantage®

Specifications

  • Description:
    Vibration Isolation Table
  • Height:
    26 - 36"
  • Cat. no.:
    82000-984
  • W×D:
    32½×25"
  • Shipping Weight:
    110 lbs.
  • Supplier No.:
    3618000

Specifications

About this item

The vibration isolation table provides an isolated work surface for conducting procedures with vibration-sensitive equipment such as microscopes and balances.

  • Four leveling feet
  • Work surface is laminated hard board

The table never makes contact with the clean bench, so vibration from the motor/blower is not transmitted to the table. Epoxy-coated steel frame, adjusts in 1" increments to provide a working height from 29.5 to 36". ADA-compliant. Compatible with Purifier Horizontal Clean Benches.

Customers Who Bought This Also Bought