Specifications
- Description:m-FC with Rosolic Acid Broth, Glass Ampules
- Size:pk/20
- Cat. no.:CA97026-198
- Supplier No.:2428520
Specifications
About this item
Selectivity: Fecal Coliform Bacteria.
-Save valuable time and money using this prepared media for membrane filtration methods.
-Broad range of media available for drinking water, wastewater, ambient water, food, and beverage applications.
-Procedures, prepared media, and required consumables available for presumptive and confirming tests.
- Save valuable time and money using this prepared media for membrane filtration methods.
- Broad range of media available for drinking water, wastewater, ambient water, food, and beverage applications.
- Procedures, prepared media, and required consumables available for presumptive and confirming tests.