White Large Display Timer, Sper Scientific
Supplier: Sper Scientific
Certificates
About this item
This easy-to-read, large display timer features the largest display available on a timer: 3×2¹/₂"
- Can be easily read from across a room
- White case has extra large start/stop button and a loud alarm
- Wall mount or fold for bench top use
- Runs for approximately two years on one AAA battery
Simultaneously displays either one count up channel and a clock, or two count down channels. All channels have a range of 24 hours with 1 second resolution. Also indicates how much time has elapsed after a count down alarm sounds and conveniently recalls the last timer setting for reuse.
Certifications: N.I.S.T. traceable certificate of compliance for certified option.
Ordering information: One AAA battery included. Five Year Warranty.
Customers Who Bought This Also Bought