Distillation Receiver, Three Flasks, Ace Glass
Catalog #: 89074-468
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Distillation receiver (complete), hose connection 5/16" tubing
- Cat. no.:89074-468
- ST Joint Size:14/20 (all)
- Supplier No.:9403-10
Specifications
About this item
This distillation receiver apparatus features three 25mL flasks that are in the same plane, rotated about the condenser axis
The hose connection is parallel to, and above, the condenser axis. Four clips are supplied for holding the flasks to condenser. Fabricated from borosilicate glass.
Ordering information: Complete item includes 89074-474 distributor; three 89074-590 flasks (25mL); and four 89059-608 Dupont Delrin joint clips/clamps. Individual components may be ordered separately.
Customers Who Bought This Also Bought