DPD Free Chlorine Reagent, SwifTest™ Dispenser, Hach
Catalog #: CA2802300
Supplier: Hach
CAS Number:
Specifications
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Description:DPD Free Chlorine Reagent, SwifTest™ Dispenser
- Size:10 mL Sample
- Method:8021 - DPD
- Compliance:EPA - YES
- Instrument:All except DR/3000, DR/2000
- Cat. no.:CA2802300
- Range:0.02 - 2.00 mg/l Cl₂
- Supplier No.:2802300
- No. of Tests:250
Specifications
About this item
DPD Free Chlorine Reagent Dispenser and Reagent Vial.
USEPA accepted for reporting drinking water analysis using Hach Method 8021.
- Range: 0.2 to 2.00 mg/L Cl2
- DPD-free
- USEPA accepted
USEPA accepted for reporting drinking water analysis using Hach Method 8021.
Fast, easy reagent addition. Just squeeze to add powder.
This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Certifications: USEPA accepted
Ordering information: Contains 1 reagent vial with enough reagent for approximately 250 Tests.
Customers Who Bought This Also Bought