Bovine;Dog;Horse;Human;Mouse;Rabbit;Rat Osteostatin (1-5) Trifluoroacetate
Catalog #: H-8645.0025BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Species:RabbitHorseHumanBovineDogRatMouse
- CAS No.:138949-73-2
- Size:25 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782492
- Protein synonyms:dog|bovine|pTH-Related Protein (107-111) (human|rat)|mouse|rabbit|Hypercalcemia of Malignancy Factor (107-111) (human|horse|pTH-rP (107-111) (human
- Protein/peptide name:Osteostatin (1-5) Trifluoroacetate
- Molecular weight:619.68
- Sequence:H-Thr-Arg-Ser-Ala-Trp-OH
- Molecular formula:C₂₇H₄₁N₉O₈
- Cat. no.:H-8645.0025BA
- Supplier No.:H-8645.0025
Specifications
About this item
The pentapeptide TRSAW, a highly conserved sequence within the pTH-related protein, is a potent inhibitor of osteoclastic bone resorption in vitro (EC₅₀ = 10⁻¹⁵ M).
Customers Who Bought This Also Bought