Ac-Lys-D-Ala-D-Ala-OH
Catalog #: H-8005.0050BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:28845-97-8
- Size:50 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00214242
- Protein/peptide name:Ac-Lys-D-Ala-D-Ala-OH
- Molecular weight:330.38
- Sequence:Ac-Lys-D-Ala-D-Ala-OH
- Molecular formula:C₁₄H₂₆N₄O₅
- Cat. no.:H-8005.0050BA
- Supplier No.:H-8005.0050
Specifications
About this item
See also F-3260.
Ac-Lys-D-Ala-D-Ala was tested as substrate in a substrate specificity study of the Streptomyces albus DD-carboxypeptidase.
Customers Who Bought This Also Bought