H-Gly-Arg-Gly-Asp-Thr-Pro-OH
Catalog #: H-7725.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:108682-58-2
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00076465
- Protein synonyms:GRGDTP
- Protein/peptide name:H-Gly-Arg-Gly-Asp-Thr-Pro-OH
- Molecular weight:601.62
- Sequence:H-Gly-Arg-Gly-Asp-Thr-Pro-OH
- Molecular formula:C₂₃H₃₉N₉O₁₀
- Cat. no.:H-7725.0005BA
- Supplier No.:H-7725.0005
Specifications
About this item
GRGDTP prevents cell attachment of fibronectin.
Customers Who Bought This Also Bought