Specifications
- CAS No.:128270-60-0
- Size:500 mg
- Storage conditions:–20 ± 5 °C
- Protein synonyms:Hirulog I|BG 8967|Hirulog
- Protein/peptide name:Bivalirudin Trifluoroacetate
- Molecular weight:2180.32
- Molecular formula:C₉₈H₁₃₈N₂₄O₃₃
- Cat. no.:H-6668.0500BA
- Supplier No.:H-6668.0500
Specifications
About this item
Highly specific and reversible direct thrombin inhibitor.
This product is offered and sold solely for uses reasonably related to the development and submission of information under a Federal law which regulates the manufacture, use or sale of drugs (the “Bolar Exemption”). Bachem cannot be made liable for any infringement of intellectual property rights. It is the sole and only responsibility of the purchaser or user of this product to comply with the relevant national rules and regulations.
Sequence: H-D-Phe-Pro-Arg-Pro-Gly-Gly-Gly-Gly-Asn-Gly-Asp-Phe-Glu-Glu-Ile-Pro-Glu-Glu-Tyr-Leu-OH
Salt: Trifluoroacetate
This product is offered and sold solely for uses reasonably related to the development and submission of information under a Federal law which regulates the manufacture, use or sale of drugs (the “Bolar Exemption”)
Bachem cannot be made liable for any infringement of intellectual property rights. It is the sole and only responsibility of the purchaser or user of this product to comply with the relevant national rules and regulations.