H-Gly-Arg-Gly-Asp-Ser-Pro-Lys-OH
Catalog #: H-6190.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:111119-28-9
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00076462
- Protein synonyms:GRGDSPK
- Protein/peptide name:H-Gly-Arg-Gly-Asp-Ser-Pro-Lys-OH
- Molecular weight:715.77
- Sequence:H-Gly-Arg-Gly-Asp-Ser-Pro-Lys-OH
- Molecular formula:C₂₈H₄₉N₁₁O₁₁
- Cat. no.:H-6190.0001BA
- Supplier No.:H-6190.0001
Specifications
About this item
GRGDSPK, fibronectin analog.
Customers Who Bought This Also Bought