Acetyl-(Pro¹⁸,Asp²¹)-beta-Amyloid (17-21) Amide
Supplier: Bachem Americas
Certificates
About this item
The pentapeptide Ac-LPFFD-NH₂ (iAβ5p) is an analog of product H-4876 with small chemical modifications which enhance its stability against proteolytic degradation. iAβ5p acts as a β-sheet breaker peptide and crosses the blood-brain barrier at a higher rate than most proteins and peptides known to be selectively taken up by the brain so that it is assumed that the peptide is being specifically transported to the brain. A significant increase in neuronal survival and decrease in brain inflammation associated with the reduction of amyloid plaques in two different transgenic Alzheimer`s disease (AD) models is additionally reported for iAβ5p.
Customers Who Bought This Also Bought