SAMS Peptide Trifluoroacetate
Catalog #: H-5938.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:125911-68-4
- Size:5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782323
- Protein synonyms:rat)|ovine)|Arg86·87)-Acetyl-CoA Carboxylase 1 (73-87) (dephosphorylated) (E. coli|(Ser77|Arg87·88)-ACC-a (74-88) (dephosphorylated) (bovine|Arg86·87)-ACC-a (73-87) (dephosphorylated) (E. coli|(Ser76|Arg87·88)-Acetyl-CoA Carboxylase 1 (74-88) (dephosphorylated) (bovine
- Protein/peptide name:SAMS Peptide Trifluoroacetate
- Molecular weight:1779.17
- Sequence:H-His-Met-Arg-Ser-Ala-Met-Ser-Gly-Leu-His-Leu-Val-Lys-Arg-Arg-OH
- Molecular formula:C₇₄H₁₃₁N₂₉O₁₈S₂
- Cat. no.:H-5938.0005BA
- Supplier No.:H-5938.0005
Specifications
About this item
Specific substrate for the sucrose-non-fermenting protein kinase (SNF1) from Saccharomyces cerevisiae and its mammalian counterpart, AMP-activated protein kinase (AMPK).
Customers Who Bought This Also Bought