(Tyr²⁷)-α-CGRP (27-37) (canine, mouse, rat)
Catalog #: H-5504.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Species:DogRatMouse
- CAS No.:124501-79-7
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00079838
- Protein/peptide name:(Tyr²⁷)-alpha-CGRP (27-37)
- Molecular weight:1182.30
- Sequence:H-Tyr-Val-Pro-Thr-Asn-Val-Gly-Ser-Glu-Ala-Phe-NH₂
- Molecular formula:C₅₄H₇₉N₁₃O₁₇
- Cat. no.:H-5504.0001BA
- Supplier No.:H-5504.0001
Specifications
About this item
Customers Who Bought This Also Bought