Human Cholecystokinin-33 Trifluoroacetate
Catalog #: H-5476.0100BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Species:Human
- CAS No.:96827-04-2
- Size:0.1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00081491
- Protein synonyms:CCK-39 (7-39)|CCK-58 (26-58)|CCK-33
- Protein/peptide name:Cholecystokinin-33 Trifluoroacetate
- Molecular weight:3945.50
- Sequence:H-Lys-Ala-Pro-Ser-Gly-Arg-Met-Ser-Ile-Val-Lys-Asn-Leu-Gln-Asn-Leu-Asp-Pro-Ser-His-Arg-Ile-Ser-Asp-Arg-Asp-Tyr(SO₃H)-Met-Gly-Trp-Met-Asp-Phe-NH₂
- Molecular formula:C₁₆₇H₂₆₃N₅₁O₅₂S₄
- Cat. no.:H-5476.0100BA
- Supplier No.:H-5476.0100
Specifications
About this item
Cholecystokinin (CCK) is a hormone originally isolated from porcine intestinal mucosa and described as a linear 33-amino acid peptide containing a sulfated tyrosine, which is essential for its biological activity. It has been found in mammals in both the digestive tract and the central nervous system. Among its multiple biological functions, this hormone stimulates pancreatic exocrine secretion, gallbladder contraction, and intestine motility and may also act as a neurotransmitter/neuromodulator in the central nervous system. For CCK-4 see H-3110.
Customers Who Bought This Also Bought