Prepro-Atrial Natriuretic Factor (56-92) Trifluoroacetate, Bachem
Catalog #: H-5474.0500BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Species:Human
- CAS No.:112199-06-1
- Size:0.5 mg
- Storage conditions:–20 ± 5 °C
- Protein synonyms:Prepro-hANF (56-92)
- Protein/peptide name:Prepro-Atrial Natriuretic Factor (56-92) Trifluoroacetate
- Molecular weight:3878.31
- Sequence:H-Glu-Val-Val-Pro-Pro-Gln-Val-Leu-Ser-Glu-Pro-Asn-Glu-Glu-Ala-Gly-Ala-Ala-Leu-Ser-Pro-Leu-Pro-Glu-Val-Pro-Pro-Trp-Thr-Gly-Glu-Val-Ser-Pro-Ala-Gln-Arg-OH
- Molecular formula:C₁₇₃H₂₇₀N₄₄O₅₇
- Cat. no.:H-5474.0500BA
- Supplier No.:H-5474.0500
Specifications
About this item
NT-pro-ANP (31-67) is used for raising antibodies.
Customers Who Bought This Also Bought