(Asp⁷⁶)-pTH (39-84) Trifluoroacetate, Bachem
Catalog #: H-4926.0500BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Species:Human
- CAS No.:79804-72-1
- Size:0.5 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD06798558
- Protein/peptide name:(Asp⁷⁶)-pTH (39-84) Trifluoroacetate
- Molecular weight:4985.55
- Sequence:H-Ala-Pro-Leu-Ala-Pro-Arg-Asp-Ala-Gly-Ser-Gln-Arg-Pro-Arg-Lys-Lys-Glu-Asp-Asn-Val-Leu-Val-Glu-Ser-His-Glu-Lys-Ser-Leu-Gly-Glu-Ala-Asp-Lys-Ala-Asp-Val-Asp-Val-Leu-Thr-Lys-Ala-Lys-Ser-Gln-OH
- Molecular formula:C₂₁₁H₃₅₆N₆₆O₇₃
- Cat. no.:H-4926.0500BA
- Supplier No.:H-4926.0500
Specifications
About this item
See also the pTH2 receptor agonist TIP39 (H-4878).
Customers Who Bought This Also Bought