E-76
Catalog #: H-4902.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- Protein/peptide name:E-76
- Molecular weight:2211.47
- Sequence:Ac-Ala-Leu-Cys-Asp-Asp-Pro-Arg-Val-Asp-Arg-Trp-Tyr-Cys-Gln-Phe-Val-Glu-Gly-NH₂
- Molecular formula:C₉₇H₁₃₉N₂₇O₂₉S₂
- Cat. no.:H-4902.0001BA
- Supplier No.:H-4902.0001
Specifications
About this item
Anticoagulant and antithrombotic peptides. Please see also the product family 'Hirudin Fragments and Analogs', the inhibitors N-1065, N-1215, N-1260, N-1565, and N-1970, and the anticoagulant anisindione, Q-1085.
The potent anticoagulant E-76, Acetyl-ALCDDPRVDRWYCQFVEG-amide, is an inhibitor of coagulation factor VIIa. It has been suggested that E-76 works by binding to an exosite on the factor VIIa protease domain, and non-competitively inhibits activation of factor X and amidolytic activity. E-76 has a well defined structure in solution determined by NMR spectroscopy that is similar to the X-ray crystal structure when complexed with factor VIIa and represents a new framework for the development of inhibitors for serine proteases.
Customers Who Bought This Also Bought