Pig PHI-27
Catalog #: H-3730.1000BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Species:Pig
- CAS No.:80458-29-3
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00133765
- Protein/peptide name:PHI-27
- Molecular weight:2995.43
- Sequence:H-His-Ala-Asp-Gly-Val-Phe-Thr-Ser-Asp-Phe-Ser-Arg-Leu-Leu-Gly-Gln-Leu-Ser-Ala-Lys-Lys-Tyr-Leu-Glu-Ser-Leu-Ile-NH₂
- Molecular formula:C₁₃₆H₂₁₆N₃₆O₄₀
- Cat. no.:H-3730.1000BA
- Supplier No.:H-3730.1000
Specifications
About this item
Porcine intestinal peptide which belongs to the glucagon-secretin family. It exhibits biological activities similar to vasoactive intestinal peptide (VIP) and secretin.
Customers Who Bought This Also Bought