H-Gly-Tyr-Gly-OH
Catalog #: H-3670.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:6099-08-7
- Size:1 g
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD00038181
- Protein/peptide name:H-Gly-Tyr-Gly-OH
- Molecular weight:295.30
- Sequence:H-Gly-Tyr-Gly-OH
- Molecular formula:C₁₃H₁₇N₃O₅
- Cat. no.:H-3670.0001BA
- Supplier No.:H-3670.0001
Specifications
About this item
GYG was used as a model peptide in studies on the UV-absorption behavior and on the nitration of Tyr-containing proteins.
Customers Who Bought This Also Bought