Galanin (1-13)-Neuropeptide Y (25-36) amide
Supplier: Bachem Americas
Certificates
About this item
The chimeric peptide M32 is a high affinity galanin receptor antagonist (IC₅₀= 0.1 nM in rat hypothalamus membranes and Kd= 0.01 nM in rat spinal cord membranes). M32 is also recognized by neuropeptide Y receptors (IC₅₀= 0.25 µM in rat cerebral cortex membranes).
Customers Who Bought This Also Bought