H-Cys-Arg-Gly-Asp-Phe-Pro-Ala-Ser-Ser-Cys-OH
Catalog #: H-2672.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:166184-23-2
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782158
- Protein synonyms:CRGDFPASSC
- Protein/peptide name:H-Cys-Arg-Gly-Asp-Phe-Pro-Ala-Ser-Ser-Cys-OH
- Molecular weight:1040.15
- Sequence:H-Cys-Arg-Gly-Asp-Phe-Pro-Ala-Ser-Ser-Cys-OH
- Molecular formula:C₄₁H₆₁N₁₃O₁₅S₂
- Cat. no.:H-2672.0001BA
- Supplier No.:H-2672.0001
Specifications
About this item
CRGDFPASSC is a cyclic RGD-containing decapeptide which binds to the fibrinogen receptor on the platelet surface. This analog with Phe substituted for Ser at position 5 was found to be 3 times more active (IC₅₀ = 2.5 µM) as a platelet aggregation inhibitor than CRGDSPASSC.
Customers Who Bought This Also Bought